(±)-3-Buten-2-ol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-Methyl vinyl carbinol, (±)-3-Hydroxy-1-butene, 1-Buten-3-ol

38707-0050 38707-0250
TS38707-0050EA 40 USD
TS38707-0050 TS38707-0250
(±)-3-Buten-2-ol 97%
(±)-3-Buten-2-ol
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 96…98 °C (760 mmHg)
Melting Pt: –100 °C
Density: 0.83 g/cm³
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00004543
CAS Number: 598-32-3
EINECS: 209-929-8
UN: 1987
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4170 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR