Diammonium-L(+)-tartrate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R,R)-(+)-Diammonium tartrate, (R,R)-(+)-Ammonium tartrate dibasic, Ammonium tartrate dibasic, L(+)-Tartaric acid diammonium salt, (+)-Diammonium L-tartrate, (R,R)-(+)-Tartaric acid diammonium salt, L(+)-Ammonium tartrate dibasic, Ammonium L-(+)-tartrate
21181-0010
21181-0050
21181-2500
TS21181-0010EA
167
USD
TS21181-0010
TS21181-0050
TS21181-2500
Diammonium-L(+)-tartrate 99%
Diammonium-L(+)-tartrate
Formula:
C₄H₁₂N₂O₆ Density: 1.6 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00013073 CAS Number: 3164-29-2 EINECS: 221-618-9 |
Specification Test Results
Appearance | Semitransparent white Crystals |
Infrared spectrum | Conforms |
Sulfated ash | ≤0.05 % |
pH | 6.0 to 7.0 (5 % solution) |
Specific optical rotation | +31.4° to +33.4° (20°C, 589 nm) (c=1.84, H2O) |
Titration with HClO4 | ≥98.5 % |
Chloride (Cl) | ≤50 ppm |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Insoluble matter | ≤0.005 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...