Oxalic acid dihydrate 99.5+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethanedioic acid dihydrate

42315-0010 42315-0051 42315-2500
TS42315-0010EA 193 USD
TS42315-0010 TS42315-0051 TS42315-2500
Oxalic acid dihydrate 99.5+% ACS
Oxalic acid dihydrate
Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
REACH: 01-2119534576-33
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Residue after ignition ≤0.01 %
Titration with KMnO4 99.5 to 102.5 %
Substance darkened by H2SO4 Passes test
Chloride (Cl) ≤0.002 %
Sulfate (SO4) ≤0.005 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤2 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR