Oxalic acid dihydrate 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

12960-0010 12960-0050 12960-1000
TS12960-0010EA 203 USD
TS12960-0010 TS12960-0050 TS12960-1000
Oxalic acid dihydrate 99+%, Extra Pure
Oxalic acid dihydrate
Formula: HO₂CCO₂H·2H₂O
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
REACH: 01-2119534576-33
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 102 °C
Sulfated ash ≤0.1 %
Titration with KMnO4 ≥99.0 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR