(±)-2-Mercaptopropionic acid 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Mercaptopropionic acid

13879-0025 13879-1000 13879-5000
TS13879-0025EA 323 USD
TS13879-0025 TS13879-1000 TS13879-5000
(±)-2-Mercaptopropionic acid 95%
(±)-2-Mercaptopropionic acid
Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 102 °C (16 mmHg)
Melting Pt: 10…14 °C
Density: 1.19 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00004862
CAS Number: 79-42-5
EINECS: 201-206-5
UN: 2936
ADR: 6.1,II
Merck Index: 12,09476

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4804 to 1.4824 (20°C, 589 nm)
Titration Iodimetric ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR