3-Mercaptopropionic acid 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
12553-0025
12553-1000
12553-5000
TS12553-0025EA
330
USD
TS12553-0025
TS12553-1000
TS12553-5000
3-Mercaptopropionic acid 99+%
3-Mercaptopropionic acid
Formula:
C₃H₆O₂S MW: 106.15 g/mol Boiling Pt: 110…111 °C (15 mmHg) Melting Pt: 17…19 °C Density: 1.218 g/cm³ Flash Pt: 124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004897 CAS Number: 107-96-0 EINECS: 203-537-0 UN: 2922 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4910 to 1.4940 (20°C, 589 nm) |
Titration Iodimetric | 99.0 to 101.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...