Potassium-D-gluconate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium Gluconate, Gluconic Acid Potassium Salt, D-Gluconic acid potassium salt, Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate

22932-0010 22932-2500
TS22932-0010EA 152 USD
TS22932-0010 TS22932-2500
Potassium-D-gluconate 99%
Potassium-D-gluconate
Formula: HOH₂C(CH(OH))₄COOK
MW: 234.25 g/mol
Melting Pt: 178…181 °C
MDL Number: MFCD00064211
CAS Number: 299-27-4
EINECS: 206-074-2
Merck Index: 12,07796

Order Now

Specification Test Results

Appearance White to off-white Crystalline or granular powder
Infrared spectrum Conforms
Assay ≥98.5 % (C6H11KO7) On dried substance
Loss on drying ≤3 % (105°C, 4 h) (Vacuum)
Heavy metals (as Pb) ≤20 ppm
Reducing substances ≤1 % (as Dextrose)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR