D-Gluconic acid 50% (w/w) in water
Supplier: Thermo Scientific Chemicals
Synonyms:
Gluconic acid, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid
11989-0010
11989-0025
11989-0100
TS11989-0010EA
51
USD
TS11989-0010
TS11989-0025
TS11989-0100
D-Gluconic acid 50% (w/w) in water
D-Gluconic acid
Formula:
C₆H₁₂O₇ MW: 196.16 Boiling Pt: 674 Melting Pt: < 15 °C Density: 1.24 Storage Temperature: Ambient |
MDL Number:
MFCD00004240 CAS Number: 526-95-4 EINECS: 208-401-4 Merck Index: 13,04469 |
Specification Test Results
Appearance | Clear yellow to brown-yellow Solution |
Titration excess NaOH | 49 to 55 % |
Reducing sugars | ≤3 % |
Titration Argentometric | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...