2-Phenylpropionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid

13087-0100 13087-0250 13087-1000
TS13087-0100EA 68 USD
TS13087-0100 TS13087-0250 TS13087-1000
2-Phenylpropionic acid 98%
2-Phenylpropionic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 260…262 °C (760 mmHg)
Melting Pt: 5 °C
Density: 1.1 g/cm³
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00002650
CAS Number: 492-37-5
EINECS: 207-752-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5210 to 1.5250 (20°C, 589 nm)
GC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR