(S)-(+)-2-Methylbutyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

22419-0010 22419-0050 22419-0250
TS22419-0010EA 130 USD
TS22419-0010 TS22419-0050 TS22419-0250
(S)-(+)-2-Methylbutyric acid 98%
(S)-(+)-2-Methylbutyric acid
Formula: C₅H₁₀O₂
Boiling Pt: 78 °C (15 mmHg)
Density: 0.938 g/cm³
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063166
CAS Number: 1730-91-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4070 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation +16° to +20° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR