Butyl butyrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyric acid butyl ester

25959-0010 25959-0050 25959-1000
TS25959-0010EA 70 USD
TS25959-0010 TS25959-0050 TS25959-1000
Butyl butyrate 98%
Butyl butyrate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: –92 °C
Density: 0.8675 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009450
CAS Number: 109-21-7
EINECS: 203-656-8
UN: 1993
ADR: 3,III
Merck Index: 13,01555

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.866 to 0.869
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR