Benzamide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzoic acid amide

15933-0025 15933-1000 15933-5000
TS15933-0025EA 319 USD
TS15933-0025 TS15933-1000 TS15933-5000
Benzamide 99%
Benzamide
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 228 °C (760 mmHg)
Melting Pt: 125…129 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00007968
CAS Number: 55-21-0
EINECS: 200-227-7
Merck Index: 13,01059

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 129 °C
HPLC ≥98.5 %
UV at 280 nm A: ≤0.05
Titration with HClO4 99.4 to 100.6 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤0.0005 %
Sodium (Na) ≤0.02 %
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR