Chloroacetic anhydride 97%

Supplier: Thermo Scientific Chemicals
Danger

40442-0250 40442-1000 40442-2500
TS40442-0250EA 58 USD
TS40442-0250 TS40442-1000 TS40442-2500
Chloroacetic anhydride 97%
Chloroacetic anhydride
Formula: C₄H₄Cl₂O₃
MW: 170.98 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 51…59 °C
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00000929
CAS Number: 541-88-8
EINECS: 208-794-2
UN: 2923
ADR: 8,II
Merck Index: 13,02130

Order Now

Specification Test Results

Appearance White to brown Powder or crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 50 to 60 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR