2-Cyanophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Cyanophenyl)metaboric acid, 2-Cyanobenzeneboronic acid, (2-Cyanophenyl)boronic acid, Dihydroxy(2-cyanophenyl)borane

36596-0010 36596-0050
TS36596-0010EA 65 USD
TS36596-0010 TS36596-0050
2-Cyanophenylboronic acid 98%
2-Cyanophenylboronic acid
Formula: C₇H₆BNO₂
MW: 146.94 g/mol
Melting Pt: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD01632208
CAS Number: 138642-62-3

Order Now

Specification Test Results

Appearance Off-white to cream Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR