1-Naphthyl boronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

34469-0010 34469-0050 34469-0250
TS34469-0010EA 28 USD
TS34469-0010 TS34469-0050 TS34469-0250
1-Naphthyl boronic acid 97%
1-Naphthyl boronic acid
Formula: C₁₀H₉BO₂
MW: 171.99 g/mol
Melting Pt: 196…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00019722
CAS Number: 13922-41-3

Order Now

Specification Test Results

Appearance Off-white to pink Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR