Adamantane 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tricyclo[3.3.1.1(3,7)]decane

14611-0010 14611-0050 14611-0250 14611-1000
TS14611-0010EA 654 USD
TS14611-0010 TS14611-0050 TS14611-0250 TS14611-1000
Adamantane 99+%
Adamantane
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Melting Pt: 205…210 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00074719
CAS Number: 281-23-2
EINECS: 206-001-4
UN: 3077
ADR: 9,III
Merck Index: 13,00149

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
GC ≥99.0 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR