Cycloheptene 95%

Supplier: Thermo Scientific Chemicals
Danger

30426-0050 30426-0250
TS30426-0050EA 107 USD
TS30426-0050 TS30426-0250
Cycloheptene 95%
Cycloheptene
Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 114.6 °C (1013 hPa)
Melting Pt: –56 °C
Density: 0.82 g/cm³
Flash Pt: –006 °C
MDL Number: MFCD00004156
CAS Number: 628-92-2
EINECS: 211-060-4
UN: 2242
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4610 (20°C, 589 nm)
GC ≥94 %
Stabilizer 0.1 % BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR