Cyclopropanecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

11162-0050 11162-0250 11162-1000
TS11162-0050EA 27 USD
TS11162-0050 TS11162-0250 TS11162-1000
Cyclopropanecarboxylic acid 98%
Cyclopropanecarboxylic acid
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: 17…19 °C
Density: 1.085 g/cm³
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001287
CAS Number: 1759-53-1
EINECS: 217-162-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4370 to 1.4390 (20°C, 589 nm)
GC ≥97.5 %
Absorbance ≤0.2 (270nm, 1g/100ml cyclohexane)
Composition 2.5 to 21.0 % (Linoleic acid)
Acid value ≤2.0 mg KOH/g
Peroxide value ≤20.0 meq O2/kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR