Cyclohexanecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexahydrobenzoic acid

14656-0010 14656-0250 14656-1000 14656-5000
TS14656-0010EA 213 USD
TS14656-0010 TS14656-0250 TS14656-1000 TS14656-5000
Cyclohexanecarboxylic acid 98%
Cyclohexanecarboxylic acid
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 30…32 °C
Density: 1.03 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00001461
CAS Number: 98-89-5
EINECS: 202-711-3

Order Now

Specification Test Results

Appearance Colorless to white Fused crystalline solid or viscous liquid
Infrared spectrum Conforms
GC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %
Elemental Analysis 9.71 to 10.21 % N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR