Cyclobutanecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

11090-0050 11090-0250 11090-1000
TS11090-0050EA 33 USD
TS11090-0050 TS11090-0250 TS11090-1000
Cyclobutanecarboxylic acid 98%
Cyclobutanecarboxylic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 195 °C (760 mmHg)
Melting Pt: –20 °C
Density: 1.04 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00001323
CAS Number: 3721-95-7
EINECS: 223-072-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4450 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR