1,2,4-Trimethylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pseudocumene

14009-0010 14009-0025 14009-0100
TS14009-0010EA 88 USD
TS14009-0010 TS14009-0025 TS14009-0100
1,2,4-Trimethylbenzene 98%
1,2,4-Trimethylbenzene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 168 °C (760 mmHg)
Melting Pt: –44 °C
Density: 0.88 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00008527
CAS Number: 95-63-6
EINECS: 202-436-9
UN: 1993
ADR: 3,III
Merck Index: 12,08100

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5050 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR