2,6-Dimethylbenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

16901-0010 16901-0100
TS16901-0010EA 37 USD
TS16901-0010 TS16901-0050 TS16901-0100
2,6-Dimethylbenzoic acid 99%
2,6-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00002483
CAS Number: 632-46-2
EINECS: 211-177-0

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 114 to 117 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR