Octylbenzene 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Phenyloctane

15099-0250 15099-1000
TS15099-0250EA 100 USD
TS15099-0250 TS15099-1000
Octylbenzene 99%
Octylbenzene
Formula: C₁₄H₂₂
MW: 190.33 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: –36 °C
Density: 0.85 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00009564
CAS Number: 2189-60-8
EINECS: 218-582-1

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4850 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR