1-Phenyl-1-propyne 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Propynylbenzene, Prop-1-yn-1-ylbenzene

17132-0050 17132-0250
TS17132-0050EA 74 USD
TS17132-0050 TS17132-0250
1-Phenyl-1-propyne 99%
1-Phenyl-1-propyne
Formula: C₆H₅CCCH₃
MW: 116.16 g/mol
Boiling Pt: 185 °C (760 mmHg)
Density: 0.92 g/cm³
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009272
CAS Number: 673-32-5
EINECS: 211-607-7

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5650 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR