Allylbenzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Phenyl-2-propene, 3-Phenylpropene
10289-0250
10289-1000
10289-5000
TS10289-0250EA
76
USD
TS10289-0250
TS10289-1000
TS10289-5000
Allylbenzene 98%
Allylbenzene
Formula:
(C₆H₅)CH₂CHCH₂ MW: 118.18 g/mol Boiling Pt: 156…157 °C (760 mmHg) Density: 0.89 g/cm³ Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008651 CAS Number: 300-57-2 EINECS: 206-095-7 UN: 3295 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5095 to 1.5135 (20°C, 589 nm) |
GC | ≥97.5 % |
Appearance of solution | Clear colorless |
Related substances | ≤0.8 % (total impurities) |
pH | 4.0 to 6.0 |
Specific optical rotation | +33.7° to +36.0° (20°C, 589 nm) (on dried substance) |
Titration with HClO4 | 99.0 to 101.0 % (on dried substance) |
Sulfates | ≤200 ppm |
Chlorides | ≤200 ppm |
Ammonium (NH4) | ≤0.1 % |
Iron (Fe) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...