2-Iodobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Iodobenzoic acid

12234-0250 12234-1000 12234-5000
TS12234-0250EA 29 USD
TS12234-0250 TS12234-1000 TS12234-5000
2-Iodobenzoic acid 98%
2-Iodobenzoic acid
Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 314
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00002419
CAS Number: 88-67-5
EINECS: 201-850-7
Merck Index: 13,05052

Order Now

Specification Test Results

Appearance White to off-white to beige Powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure
Calcium (Ca) ≤0.003 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.01 %
Insoluble matter ≤0.005 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR