Trimellitic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,4-Benzenetricarboxylic acid, Benzene-1,2,4-tricarboxylic acid

34903-0050 34903-0500 34903-2500
TS34903-0050EA 29 USD
TS34903-0050 TS34903-0500 TS34903-2500
Trimellitic acid 98%
Trimellitic acid
Formula: C₉H₆O₆
MW: 210.14 g/mol
Melting Pt: 231 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002470
CAS Number: 528-44-9
EINECS: 208-432-3
Merck Index: 12,09832

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR