2,5-Dihydroxybenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
16520-0050
16520-0500
16520-2500
TS16520-0050EA
26
USD
TS16520-0050
TS16520-0500
TS16520-2500
2,5-Dihydroxybenzoic acid 99%
2,5-Dihydroxybenzoic acid
Formula:
C₇H₆O₄ Melting Pt: 204…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002460 CAS Number: 490-79-9 EINECS: 207-718-5 Merck Index: 13,04409 |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 204 to 207 °C |
HPLC | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...