Salicylic acid 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid

14770-0010 14770-0025
TS14770-0010EA 74 USD
TS14770-0010 TS14770-0025
Salicylic acid 99+%
Salicylic acid
Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Boiling Pt: 211 °C (20 mmHg)
Melting Pt: 158…161 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 13,08411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1f695fa92656a135b5e1742f1cca28f4d255fb&vid=8f3aff6fb9a70e5dd194b8bc46019af74e48e90a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 158 to 161 °C
Titration with NaOH 99.0 to 101.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR