3,5-Dimethoxybenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

11546-0050 11546-0250 11546-1000
TS11546-0050EA 25 USD
TS11546-0050 TS11546-0250 TS11546-1000
3,5-Dimethoxybenzoic acid 99%
3,5-Dimethoxybenzoic acid
Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 341 °C (1013 hPa)
Melting Pt: 182…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00002502
CAS Number: 1132-21-4
EINECS: 214-473-8

Order Now

Specification Test Results

Appearance White to pink-beige Crystalline powder
Infrared spectrum Conforms
Melting point 182 to 185 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR