3,4-Dimethoxybenzoic acid 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Veratric acid
11545-0250
11545-1000
11545-5000
TS11545-0250EA
28
USD
TS11545-0250
TS11545-1000
TS11545-5000
3,4-Dimethoxybenzoic acid 99+%
3,4-Dimethoxybenzoic acid
Formula:
C₉H₁₀O₄ Melting Pt: 179…182 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002500 CAS Number: 93-07-2 EINECS: 202-215-7 Merck Index: 12,10086 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 180 to 184 °C |
HPLC | ≥99.0 % |
Color scale | ≤10 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...