3,5-Dinitrobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

15319-0010 15319-1000 15319-5000
TS15319-0010EA 175 USD
TS15319-0010 TS15319-1000 TS15319-5000
3,5-Dinitrobenzoic acid 98+%
3,5-Dinitrobenzoic acid
Formula: C₇H₄N₂O₆
Melting Pt: 205…209 °C
Density: 1.683 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007253
CAS Number: 99-34-3
EINECS: 202-751-1
Merck Index: 13,03304

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 204 to 209 °C
HPLC ≥98.0 %
Titration with NaOH 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR