Benzoic acid 99.5% for analysis

Supplier: Thermo Scientific Chemicals
Danger

22180-0010 22180-0050 22180-2500
TS22180-0010EA 182 USD
TS22180-0010 TS22180-0050 TS22180-2500
Benzoic acid 99.5% for analysis
Benzoic acid
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 249 °C (760 mmHg)
Melting Pt: 121…123 °C
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 121 to 123 °C
Sulfated ash ≤0.01 %
Titration with NaOH ≥99.5 %
Sulfate (SO4) ≤30 ppm
Iron (Fe) ≤4 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR