Benzoic acid 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
14913-0010
14913-0025
14913-0250
14913-5000
TS14913-0010EA
73
USD
TS14913-0010
TS14913-0025
TS14913-0250
TS14913-5000
Benzoic acid 99%, Extra Pure
Benzoic acid
Formula:
C₇H₆O₂ Boiling Pt: 249 °C (760 mmHg) Melting Pt: 121…123 °C Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002398 CAS Number: 65-85-0 EINECS: 200-618-2 REACH: 01-2119455536-33 Merck Index: 13,01092 |
|
Specification Test Results
Appearance | White to off-white Flakes |
Infrared spectrum | Conforms |
Melting point | 121 to 124 °C |
Water | ≤0.5 % (K.F.) |
Sulfated ash | ≤0.01 % |
Titration with NaOH | ≥98.5 % |
Clarity of alcohol solution | (20 wt.% soln. in ethanol) clear |
Color of molten substance | ≤50 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...