Benzoic acid methyl ester 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl benzoate

12634-0025 12634-0100 12634-5000
TS12634-0025EA 169 USD
TS12634-0025 TS12634-0100 TS12634-5000
Benzoic acid methyl ester 99%
Benzoic acid methyl ester
Formula: C₆H₅COOCH₃
MW: 136.15 g/mol
Boiling Pt: 199.6 °C (1013 hPa)
Melting Pt: –12 °C
Density: 1.08 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00008421
CAS Number: 93-58-3
EINECS: 202-259-7
Merck Index: 13,06052
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=287e16081e44f5525ff8b4d34f0a6fece44a3073&vid=2be9af3b90e55af477c374c091cd0888dcf60b96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5180 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25/25°C) 1.084 to 1.088
Acid value ≤1 mg KOH/g
UV at 245 nm A: ≤1.00
Color scale ≤10 APHA
Stabilizer 0.5 to 1.0 % Ethanol
Residue after evaporation ≤0.0002 %
Free chlorine ≤0.00003 % (as Cl)
Free acid (HCl) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR