4-Octyne 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dipropylacetylene

22352-0100 22352-0500
TS22352-0100EA 52 USD
TS22352-0100 TS22352-0500
4-Octyne 98+%
4-Octyne
Formula: C₈H₁₄
MW: 110.2 g/mol
Boiling Pt: 131…132 °C (760 mmHg)
Melting Pt: –103 °C
Density: 0.75 g/cm³
Flash Pt: 29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009471
CAS Number: 1942-45-6
EINECS: 217-730-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4225 to 1.4275 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR