2-Butyne 98%

Supplier: Thermo Scientific Chemicals
Danger

13514-0010 13514-0050
TS13514-0010EA 29 USD
TS13514-0010 TS13514-0050
2-Butyne 98%
2-Butyne
Formula: C₄H₆
Boiling Pt: 27 °C (1013 hPa)
Melting Pt: –32 °C
Density: 0.69 g/cm³
Flash Pt: –25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009275
CAS Number: 503-17-3
EINECS: 207-962-2
UN: 1144
ADR: 3,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3920 to 1.3940 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR