2,4,4-Trimethyl-2-pentene 98%

Supplier: Thermo Scientific Chemicals
Danger

16263-0050 16263-1000
TS16263-0050EA 67 USD
TS16263-0050 TS16263-1000
2,4,4-Trimethyl-2-pentene 98%
2,4,4-Trimethyl-2-pentene
Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: –106 °C
Density: 0.72 g/cm³
Flash Pt: 2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008902
CAS Number: 107-40-4
EINECS: 203-486-4
UN: 2050
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4170 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR