2,4,4-Trimethyl-1-pentene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14018-1000
14018-5000
TS14018-1000EA
128
USD
TS14018-1000
TS14018-5000
2,4,4-Trimethyl-1-pentene 99%
2,4,4-Trimethyl-1-pentene
Formula:
C₈H₁₆ Boiling Pt: 101…102 °C (760 mmHg) Melting Pt: –101 °C Density: 0.7 g/cm³ Flash Pt: –6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008855 CAS Number: 107-39-1 EINECS: 203-486-4 UN: 2050 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4070 to 1.4090 (20°C, 589 nm) |
GC | ≥98.5 % |
Specific optical rotation | +25° to +30° (20°C, 589 nm) (c=1 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...