2,3-Dimethyl-2-butene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetramethylethylene

15659-0250 15659-1000 15659-5000
TS15659-0250EA 89 USD
TS15659-0250 TS15659-1000 TS15659-5000
2,3-Dimethyl-2-butene 98%
2,3-Dimethyl-2-butene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 73 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.7 g/cm³
Flash Pt: –16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008897
CAS Number: 563-79-1
EINECS: 209-263-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4130 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR