2,3,3-Trimethyl-1-butene 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14013-0050
14013-0250
TS14013-0050EA
98
USD
TS14013-0050
TS14013-0250
2,3,3-Trimethyl-1-butene 99+%
2,3,3-Trimethyl-1-butene
Formula:
C₇H₁₄ MW: 98.19 g/mol Boiling Pt: 78 Melting Pt: -109.9 Density: 0.7 g/cm³ Flash Pt: –17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008850 CAS Number: 594-56-9 EINECS: 209-845-1 UN: 3295 ADR: 3 II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4015 to 1.4035 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...