trans-3-Hexene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (E)-3-Hexene, 3-Hexene

14991-0010 14991-0051
TS14991-0010EA 45 USD
TS14991-0010 TS14991-0051
trans-3-Hexene 98%
trans-3-Hexene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 67 °C (760 mmHg)
Melting Pt: –113 °C
Density: 0.677 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00009386
CAS Number: 13269-52-8
EINECS: 236-261-4
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3935 to 1.3955 (20°C, 589 nm)
Assay ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR