trans-2-Hexene 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (E)-2-Hexene

14990-0050 14990-0250
TS14990-0050EA 115 USD
TS14990-0050 TS14990-0250
trans-2-Hexene 98+%
trans-2-Hexene
Formula: C₆H₁₂
Boiling Pt: 68 °C (1013 hPa)
Melting Pt: –98.5 °C
Density: 0.66 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00009473
CAS Number: 4050-45-7
EINECS: 223-752-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3920 to 1.3940 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR