cis-2-Pentene

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Z)-2-Pentene

38359-0010 38359-0050 38359-0100
TS38359-0010EA 37 USD
TS38359-0010 TS38359-0050 TS38359-0100
cis-2-Pentene
cis-2-Pentene
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 37 °C (760 mmHg)
Melting Pt: –151 °C
Density: 0.65 g/cm³
Flash Pt: –27 °C
Storage Temperature: Ambient
MDL Number: MFCD00063978
CAS Number: 627-20-3
EINECS: 210-988-7
UN: 3295
ADR: 3,II
Merck Index: 12,07263

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3810 to 1.3830 (20°C, 589 nm)
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR