1-Pentene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: alpha-n-Amylene, Propylethylene

26866-0025 26866-0250 26866-1000 26866-5000
TS26866-0025EA 690 USD
TS26866-0025 TS26866-0250 TS26866-1000 TS26866-5000
1-Pentene 97%
1-Pentene
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 30 °C (760 mmHg)
Melting Pt: –165 °C
Density: 0.64 g/cm³
Flash Pt: –28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003567
CAS Number: 109-67-1
EINECS: 203-694-5
UN: 1108
ADR: 3,I
Merck Index: 12,07262

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3704 to 1.3724 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR