1-Octene 99+%

Supplier: Thermo Scientific Chemicals
Danger

30125-0010 30125-0025 30125-0100 30125-1000 30125-5000
TS30125-0010EA 84 USD
TS30125-0010 TS30125-0025 TS30125-0100 TS30125-1000 TS30125-5000
1-Octene 99+%
1-Octene
Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 121…123 °C (760 mmHg)
Melting Pt: –102 °C
Density: 0.715 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00009548
CAS Number: 111-66-0
EINECS: 203-893-7
UN: 3295
ADR: 3 II
Merck Index: 13,01770

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4100 (20°C, 589 nm)
Water ≤0.005 %
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR