1-Heptene 98%

Supplier: Thermo Scientific Chemicals
Danger

12039-0250 12039-1000
TS12039-0250EA 112 USD
TS12039-0250 TS12039-1000
1-Heptene 98%
1-Heptene
Formula: C₇H₁₄
MW: 98.19 g/mol
Boiling Pt: 94 °C (760 mmHg)
Melting Pt: –119 °C
Density: 0.697 g/cm³
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00009531
CAS Number: 592-76-7
EINECS: 209-767-8
UN: 2278
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3990 to 1.4010 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR