1-Dodecene 93-95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dodec-1-ene
11763-0025
11763-0100
11763-1000
11763-5000
TS11763-0025EA
352
USD
TS11763-0025
TS11763-0100
TS11763-1000
TS11763-5000
1-Dodecene 93-95%
1-Dodecene
Formula:
C₁₂H₂₄ MW: 168.32 g/mol Boiling Pt: 213 °C (1013 hPa) Melting Pt: –35 °C Density: 0.75 g/cm³ Flash Pt: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008961 CAS Number: 112-41-4 EINECS: 203-968-4 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4284 to 1.4304 (20°C, 589 nm) |
GC | ≥99.8 % |
Solubility | (In water) Passes test |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Substance darkened by H2SO4 | Passes test |
Carbonyl compounds | Each of acetone, formaldehyde and acetaldehyde |
Titrable acid | ≤0.0003 meq/g |
Titrable base | ≤0.0002 meq/g |
KMnO4-reducing substance | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...