1-Decene ca. 95%

Supplier: Thermo Scientific Chemicals
Danger

11191-0010 11191-0025
TS11191-0010EA 63 USD
TS11191-0010 TS11191-0025
1-Decene ca. 95%
1-Decene
Formula: C₁₀H₂₀
Boiling Pt: 169…171 °C
Melting Pt: –66 °C
Density: 0.741 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009577
CAS Number: 872-05-9
EINECS: 212-819-2
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20°C, 589 nm)
GC ≥94 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR