Isopropenyl acetate 99%

Supplier: Thermo Scientific Chemicals
Danger

15250-0010 15250-2500
TS15250-0010EA 84 USD
TS15250-0010 TS15250-2500
Isopropenyl acetate 99%
Isopropenyl acetate
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 94 °C (1013 hPa)
Melting Pt: –93 °C
Density: 0.92 g/cm³
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00008709
CAS Number: 108-22-5
EINECS: 203-562-7
UN: 2403
ADR: 3,II
Merck Index: 13,05223

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4000 to 1.4020 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR